missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE1L3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11330-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the DNase family. The protein hydrolyzes DNA, is not inhibited by actin, and mediates the breakdown of DNA during apoptosis. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Spécification
DNASE1L3 | |
Polyclonal | |
Unconjugated | |
DNASE1L3 | |
Deoxyribonuclease gamma, deoxyribonuclease I like 3, DHP2, DNAS1L3, DNase gamma, DNase I homolog protein DHP2, DNase I like 3, DNASE1L3, Liver and spleen DNase, LS DNase, LSD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116687, 13421, 1776 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O55070, O89107, Q13609 | |
DNASE1L3 | |
DNASE1L3 Fusion Protein Ag1873 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |