missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27151-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
SRC2.Spécification
DOCK4 | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N1I0 | |
dedicator of cytokinesis 4, DOCK4, KIAA0716 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
9732 |
Western Blot | |
Unconjugated | |
DOCK4 | |
Liquid | |
DOCK4 | |
DOCK4 Fusion Protein Ag25946 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |