Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Qdot™ 625, Invitrogen™
Donkey Polyclonal Secondary Antibody
Marque:  Invitrogen Q22084
Code produit. 12303643
  • 319.00€ / 1 pièce
Découvrez plus d'offres spéciales
Description
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Goat IgG (H+L) Polyclonal antibody specifically detects Goat IgG (H+L) in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Goat IgG (H+L)
Polyclonal
Qdot 625
Donkey
purified
RUO
Goat
Liquid
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
1 μM
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3
Gamma Immunoglobins Heavy and Light chains
100 μL
Secondary
4° C, store in dark