Learn More
Tocris Bioscience™ DOPE
Click to view available options
Quantité:
25 mg
Conditionnement:
25mg
Description
DOPE is a liposome/lipoplex transfection reagent. Enhances liposome and lipoplex transfection efficiency in combination with DA, DOTAP or DC-cholesterol (Cat. No. 7176). Please note: exact ratio for use and transfection efficiency is dependent on cell line used.
Spécification
Spécification
| Nom chimique ou matériau | (9 Z)-1,1 '-[(1 R)-1-[[[(2-Aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl] 9-octadecenoic acid ester |
| CAS | 4004-05-1. |
| Quantité | 25 mg |
| Cible | General Laboratory Research Reagent |
| Formule moléculaire | C41H78NO8P |
| Pureté | 0.95 |