Learn More
Dowex™ 1X8-400, résine échangeuse d’ions, Thermo Scientific Chemicals
Catalog Number p-2885573
Click to view available options
Quantité:
100 g
500 g
2.5 kg
Conditionnement:
Flacon en plastique
12627-85-9
C29H34ClN
432.05
MFCD00132718
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
*
12627-85-9
C29H34ClN
432.05
MFCD00132718
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
*
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Dowex™ 1X8
Forme physique Microbilles humides
Granulométrie 15 % max. (on 100 mesh, cumulative) 15 % max. (through 325 mesh)
Conditionnement Flacon en plastique
Capacité totale de volume wet volume capacity, Cl form (meq/mL):> 1.2
Capacité de rétention d’eau Cl form: 39 to 45%
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Couleur Blanc à jaune
Merck Index 10, 2182
Quantité 100 g
Nom IUPAC 1,4-bis(éthényl)benzène ; (4-éthénylphényl)-triméthylazanium ; styrène ; chlorure