Learn More
DRP1 (N-terminal) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 26187-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16804715
  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
Description
Dynamin-related protein 1 (DRP1) is a member of the dynamin superfamily of GTPases. Members of this family have diverse cellular functions including vesicle scission, organelle fission, viral resistance, and intracellular trafficking (reviewed in 1). DRP1 affects mitochondrial morphology and is important in mitochondrial and peroxisomal fission in mammalian cells.
Spécification
Spécification
DRP1 (N-terminal)
Polyclonal
Unconjugated
DNM1L
DLP1, DNM1L, DRP1
Rabbit
Antigen Affinity Chromatography
RUO
10059, 74006
-20°C
Liquid
Western Blot, Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00429, Q8K1M6
Dnm1l
DRP1 (N-terminal) Fusion Protein Ag24249
20 μL
Primary
Mouse, Human