Learn More
DUOX1 Mouse anti-Human, Unconjugated, Clone: 3B8D6, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67226-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The DUOX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DUOX1. This antibody recognizes human antigen. The DUOX1 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
DUOX1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
3B8D6 | |
Unconjugated | |
150 μL | |
53905 | |
IgG2a |