missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DUX4 (Human) Recombinant Protein

Human DUX4 full-length ORF (AAI60122.1, 1 a.a. - 424 a.a.) recombinant protein with GST tag at N-terminal.

Marque:  Abnova™ H00022947-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16126792

  • 295.00€ / 10µg

Expédition estimée
04-01-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

  • Sequence: MALPTPSDSTLPAEARGRGRRRRLVWTPSQSEALRACFERNPYPGIATRERLAQAIGIPEPRVQIWFQNERSRQLRQHRRESRPWPGRRGPPEGRRKRTAVTGSQTALLLRAFEKDRFPGIAAREELARETGLPESRIQIWFQNRRARHPGQGGRAPAQAGGLCSAAPGGGHPAPSWVAFAHTGAWGTGLPAPHVPCAPGALPQGAFVSQAARAAPALQPSQAAPAEGVSQPAPARGDFAYAAPAPPDGALSHPQAPRWPPHPGKSREDRDPQRDGLPGPCAVAQPGPAQAGPQGQGVLAPPTSQGSPWWGWGRGPQVAGAAWEPQAGAAPPPQPAPPDASASARQGQMQGIPAPSQALQEPAPWSALPCGLLLDELLASPEFLQQAQPLLETEAPGELEASEEAASLEAPLSEEEYRALLEEL
Spécification

Spécification

AAI60122.1
double homeobox, 4
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DUX4
GST
22947
Wheat germ expression system
10 μg
DUX10
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Vidéos
FDS
Documents