missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27615-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F2, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner.Spécification
E2F3 | |
Polyclonal | |
Unconjugated | |
E2F3 | |
E2F 3, E2F transcription factor 3, E2F3, KIAA0075, Transcription factor E2F3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1871 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00716 | |
E2F3 | |
E2F3 Fusion Protein Ag26367 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |