Learn More
STEMCELL Technologies Easy 50 EasySep™ Magnet
Immunomagnetic column-free magnet
Marque: STEMCELL Technologies 18002
Informations supplémentaires : Poids : 0.05000kg
Description
- The Easy 50 EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a large number of cells (up to 4 x 10^9 cells)
- The Easy 50 EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with magnetic EasySep™ particles without the use of columns
- The Easy 50 EasySep™ Magnet is designed to hold a standard 50 mL conical tube
Spécification
4 x 109 cells | |
Cell Isolation |