missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDEM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27310-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Involved in endoplasmic reticulum-associated degradation (ERAD). Accelerates the glycoprotein ERAD by proteasomes, by catalyzing mannose trimming from Man8GlcNAc2 to Man7GlcNAc2 in the N-glycans. Seems to have alpha 1,2-mannosidase activity.Spécification
EDEM3 | |
Polyclonal | |
Unconjugated | |
EDEM3 | |
C1orf22, EDEM3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66967, 80267 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2HXL6, Q9BZQ6 | |
Edem3 | |
EDEM3 Fusion Protein Ag26123 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |