missing translation for 'onlineSavingsMsg'
Learn More

EHF Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27195-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805475

  • 112.71€ / 20µL

Expédition estimée
25-02-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein that belongs to an ETS transcription factor subfamily characterized by epithelial-specific expression (ESEs). The encoded protein acts as a transcriptional repressor and may be associated with asthma susceptibility. This protein may be involved in epithelial differentiation and carcinogenesis.
Spécification

Spécification

EHF
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NZC4
EHF, ESE 3, ESE3, ESE3B, ESEJ, ets homologous factor, hEHF
Rabbit
IgG
20 μL
-20°C
Primary
26298
Western Blot
Unconjugated
EHF
Liquid
EHF
EHF Fusion Protein Ag25885
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents