missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2AK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20499-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The HRI gene is localized to 7p22 where its 3′ end slightly overlaps the 3′ end of the gene JTV1. The two genes are transcribed from opposite strands. Studies in rat and rabbit suggest that the HRI gene product phosphorylates the alpha subunit of eukaryotic initiation factor 2. Its kinase activity is induced by low levels of heme and inhibited by the presence of heme.Spécification
EIF2AK1 | |
Polyclonal | |
Unconjugated | |
EIF2AK1 | |
EIF2AK1, HCR, Heme controlled repressor, Heme regulated inhibitor, HRI, KIAA1369 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15467, 27102, 27137 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q63185, Q9BQI3, Q9Z2R9 | |
Eif2ak1 | |
EIF2AK1 Fusion Protein Ag14236 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |