missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELAC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10071-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3′ trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
ELAC2 | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80Y81, Q8CGS5, Q9BQ52 | |
elaC homolog 2 (E. coli), ElaC homolog protein 2, ELAC2, ELC2, HPC2, Ribonuclease Z 2, RNase Z 2, tRNA 3 endonuclease 2, tRNase Z 2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
282826, 60528, 68626 |
Western Blot | |
Unconjugated | |
ELAC2 | |
Liquid | |
Elac2 | |
ELAC2 Fusion Protein Ag0105 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |