Learn More
Merck™ Purification Kit
With ion-exchange technology
Marque:  Merck™ QGARD00D3
Code nomenclature Nacres: JA.13
Informations supplémentaires : Poids : 0.50000kg
Code produit. 10001592
  • 465.00€ / 1 pièce
Expédition estimée
15-05-2019
Ajouter au panier
Description
Description
Q-Gard™ 3 Purification Cartridge for Milli-Q™ systems connected to DI water feed containing high silt
Spécification
Spécification
Q-Gard 3
Purification Cartridge
Depth Filtration, Ion-Exchange, Activated Carbon
Deionized
Milli-Q™systems connected to DI water feed containing high silt
Vidéos