missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entactin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | Entactin |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16812154
|
Proteintech
13766-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16802154
|
Proteintech
13766-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Although the precise composition and structure of basal laminae varies from tissue to tissue, as well as within the same tissue at different developmental stages, most mature basal laminae contain type IV collagen, laminins, heparan sulphate proteoglycans and entactin (or nidogen). Entactin is a sulfated glycoprotein that is widely distributed in basement membranes. It is tightly associated with laminin and also binds to collagen IV. Entactin probably has a role in cell-extracellular matrix interactions.Spécification
Entactin | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P08460, P10493, P14543 | |
18073, 25494, 4811 | |
Entactin Fusion Protein Ag4749 | |
Primary | |
-20°C | |
NID1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Entactin, NID, NID 1, NID1, Nidogen 1 | |
Nid1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |