missing translation for 'onlineSavingsMsg'
Learn More

Entactin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13766-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802154

  • 329.85€ / 150µL

Expédition estimée
26-02-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Although the precise composition and structure of basal laminae varies from tissue to tissue, as well as within the same tissue at different developmental stages, most mature basal laminae contain type IV collagen, laminins, heparan sulphate proteoglycans and entactin (or nidogen). Entactin is a sulfated glycoprotein that is widely distributed in basement membranes. It is tightly associated with laminin and also binds to collagen IV. Entactin probably has a role in cell-extracellular matrix interactions.
Spécification

Spécification

Entactin
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08460, P10493, P14543
Entactin, NID, NID 1, NID1, Nidogen 1
Rabbit
IgG
150 μL
-20°C
Primary
18073, 25494, 4811
Immunoprecipitation, Western Blot
Unconjugated
NID1
Liquid
Nid1
Entactin Fusion Protein Ag4749
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents