missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15778-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
ENY2 is a component of the transcription regulatory histone acetylation (HAT) complex SAGA, a multiprotein complex that activates transcription by remodeling chromatin and mediating histone acetylation and deubiquitination. Within the SAGA complex, participates in a subcomplex that specifically deubiquitinates both histones H2A and H2B. The SAGA complex is recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation. May also participate in mRNA export and accurate chromatin positioning in the nucleus by tethering genes to the nuclear periphery.Spécification
ENY2 | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JIX0, Q9NPA8 | |
DC6, e(y)2, ENY2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
223527, 56943, 685258 |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
ENY2 | |
Liquid | |
ENY2 | |
ENY2 Fusion Protein Ag8462 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |