missing translation for 'onlineSavingsMsg'
Learn More

EOMES Monoclonal Antibody (Dan11mag), PE-Cyanine5, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 15-4875-82

321.34 EUR valable jusqu'au 2024-03-29
Utilisez le code promo "21615" pour bénéficier de cette offre.



Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16370654

  • 482.00€ / 100µg
En stock
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Rat Monoclonal Antibody

TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
Spécification

Spécification

EOMES
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O54839
Eomes
Affinity chromatography
RUO
13813
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
Dan11mag
PE-Cyanine5
Eomes
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions