missing translation for 'onlineSavingsMsg'
Learn More

Éthyl 3-trifluorométhyl-1H-pyrazole-4-carboxylate, 97 %, Thermo Scientific Chemicals

Catalog Number p-7021324
Click to view available options
Quantité:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
155377-19-8
C7H7F3N2O2
208.14
MFCD00052083
VYXIHSAEOXPAEY-UHFFFAOYSA-N
ethyl 3-trifluoromethyl pyrazole-4-carboxylate, ethyl 3-trifluoromethyl-1h-pyrazole-4-carboxylate, ethyl 5-trifluoromethyl-1h-pyrazole-4-carboxylate, 3-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-trifluoromethyl pyrazole-4-carboxylic acid ethyl ester, ethyl 3-trifluoromethyl-2h-pyrazole-4-carboxylate, 5-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 1h-pyrazole-4-carboxylic acid, 3-trifluoromethyl-, ethyl ester
596095
5-(trifluorométhyl)-1H-pyrazole-4-carboxylate d’éthyle
CCOC(=O)C1=C(NN=C1)C(F)(F)F
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
155377-19-8
C7H7F3N2O2
208.14
MFCD00052083
VYXIHSAEOXPAEY-UHFFFAOYSA-N
ethyl 3-trifluoromethyl pyrazole-4-carboxylate, ethyl 3-trifluoromethyl-1h-pyrazole-4-carboxylate, ethyl 5-trifluoromethyl-1h-pyrazole-4-carboxylate, 3-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-trifluoromethyl pyrazole-4-carboxylic acid ethyl ester, ethyl 3-trifluoromethyl-2h-pyrazole-4-carboxylate, 5-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 1h-pyrazole-4-carboxylic acid, 3-trifluoromethyl-, ethyl ester
596095
5-(trifluorométhyl)-1H-pyrazole-4-carboxylate d’éthyle
CCOC(=O)C1=C(NN=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate
Point de fusion 142°C to 146°C
Quantité 1 g
Poids de la formule 208.14
Pourcentage de pureté 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'