missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24295-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.Spécification
F2 | |
Polyclonal | |
Unconjugated | |
F2 | |
Coagulation factor II, F2, Prothrombin, PT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2147 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00734 | |
F2 | |
F2 Fusion Protein Ag18385 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |