missing translation for 'onlineSavingsMsg'
Learn More

FAM126A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26243-1-AP-20UL

Additional Details : Poids : 0.00010kg

 View more versions of this product

Product Code. 16804765

  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.
Specifications

Specifications

FAM126A
Polyclonal
Unconjugated
FAM126A
DRCTNNB1A, FAM126A, HCC, HLD5, HYCC1, Hyccin, Protein FAM126A
Rabbit
Antigen Affinity Chromatography
RUO
84668
-20°C
Liquid
Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BYI3
FAM126A
FAM126A Fusion Protein Ag24004
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers