missing translation for 'onlineSavingsMsg'
Learn More

FBLIM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13349-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16801514

  • 112.71€ / 20µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is a rabbit polyclonal antibody raised against full length human FBLIM1 antigen.

This gene encodes a protein with an N-terminal filamin-binding domain, a central proline-rich domain, and, multiple C-terminal LIM domains. This protein localizes at cell junctions and may link cell adhesion structures to the actin cytoskeleton. This protein may be involved in the assembly and stabilization of actin-filaments and likely plays a role in modulating cell adhesion, cell morphology and cell motility. This protein also localizes to the nucleus and may affect cardiomyocyte differentiation after binding with the CSX/NKX2-5 transcription factor. Alternative splicing results in multiple transcript variants encoding different isoforms.
Spécification

Spécification

FBLIM1
0.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q71FD7, Q8WUP2
CAL, FBLIM1, FBLP 1, FBLP1, filamin binding LIM protein 1, MIG2 interacting protein, Migfilin, RP11 169K16.5
Rabbit
IgG
20 μL
-20°C
Primary
362650, 54751, 74202
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FBLIM1
Liquid
Fblim1
FBLIM1 Fusion Protein Ag3907
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Vidéos
FDS
Documents