missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13934-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
FBXL14 is members of the F-box protein family, such as FBXL14, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.Spécification
FBXL14 | |
Polyclonal | |
Unconjugated | |
FBXL14 | |
F box/LRR repeat protein 14, Fbl14, FBXL14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101358, 144699, 312675 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BID8, Q8N1E6 | |
Fbxl14 | |
FBXL14 Fusion Protein Ag4975 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |