missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCER1A Armenian Hamster, FITC, Clone: 42795, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marque: EnquireBio QAB91-F-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
FCER1A | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
FCER1A | |
Protein A/G Purified | |
RUO | |
2205 |
Flow Cytometry | |
42795 | |
FITC | |
NP_001992 | |
Armenian Hamster | |
25 μg | |
Primary | |
IgG |