missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27100-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Fibroblast growth factors (FGFs) can stimulate the proliferation of mesenchymal, epithelial and neuroectodermal cells. FGF3 belongs to the heparin binding growth factors family. Members of the family include Acidic FGF (also known as aFGF or FGF 1), basic FGF (bFGF or FGF 2), FGF 3 (also known as Int2) and FGF 4 (also known as hst or Kaposi FGF). FGF 4 was first revealed to be an oncoprotein in Kaposi′s sarcoma cells. Other family members include FGF 5, FGF 6, FGF 7 (keratinocyte growth factor, KGF), FGF 8 (AIGF), FGF 9 (GAF) and FGF 10. FGF3 is activated by proviral insertion of mouse mammary tumor virus (MMTV). FGF3 is responsible for most of breast maligcies.Spécification
FGF3 | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11487 | |
FGF 3, FGF3, Fibroblast growth factor 3, HBGF 3, INT2, Proto oncogene Int 2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
2248 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
FGF3 | |
Liquid | |
FGF3 | |
FGF3 Fusion Protein Ag25361 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |