missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FMNL2 Recombinant Protein
Marque: Abnova™ H00114793-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
AAH36492 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
45.32 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MDLTKREYTMHDHNTLLKEFILNNEGKLKKLQDDAKIAQDAFDDVVKYFGENPKTTPPSVFFPVFVRFVKAYKQAEEENELRKKQEQALMEKLLEQEALMEQQDPKSPSHKSKRQQQELIAELRRRQVKDNRHVYEGKDGAIEDIITALKKNNITKFPNVHSRVRISSSTPVVEDTQS | |
FHOD2/FLJ37546 | |
FMNL2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
114793 | |
FMNL2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FMNL2 | |
Human | |
Recombinant | |
Solution |