missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Foxp3 Mouse, APC, Clone: 3G3, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio QAB71-APC-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Foxp3 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH7.2 | |
Foxp3 | |
Protein A/G Purified | |
RUO | |
20371 | |
IgG1 κ |
Flow Cytometry | |
3G3 | |
APC | |
NP_001186276 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-4°C. |