missing translation for 'onlineSavingsMsg'
Learn More

FOXQ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23718-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802425

  • 112.71€ / 20µL

Expédition estimée
25-02-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
Spécification

Spécification

FOXQ1
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70220, Q63244, Q9C009
Forkhead box protein Q1, forkhead box Q1, FOXQ1, HFH 1, HFH1, HNF 3/forkhead like protein 1
Rabbit
IgG
20 μL
-20°C
Primary
15220, 64826, 94234
Western Blot
Unconjugated
FOXQ1
Liquid
Foxq1
FOXQ1 Fusion Protein Ag20545
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Vidéos
FDS
Documents