missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTSJD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27707-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Sodium-phosphate symporter which seems to play a fundamental housekeeping role in phosphate transport by absorbing phosphate from interstitial fluid for normal cellular functions such as cellular metabolism, signal transduction, and nucleic acid and lipid synthesis. In vitro, sodium-dependent phosphate uptake is not siginificantly affected by acidic and alkaline conditions, however sodium-independent phosphate uptake occurs at acidic conditions. May play a role in extracellular matrix, cartilage and vascular calcification. Functions as a retroviral receptor and confers human cells susceptibility to infection to amphotropic murine leukemia virus (A-MuLV), 10A1 murine leukemia virus (10A1 MLV) and some feline leukemia virus subgroup B (FeLV-B) variants.Spécification
FTSJD2 | |
Polyclonal | |
Unconjugated | |
CMTR1 | |
CMTR1, FTSJD2, hMTr1, ISG95, KIAA0082, MTr1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23070, 74157 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N1G2, Q9DBC3 | |
Cmtr1 | |
FTSJD2 Fusion Protein Ag26722 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |