missing translation for 'onlineSavingsMsg'
Learn More

FUBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24864-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16803365

  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase.
Spécification

Spécification

FUBP1
Polyclonal
Unconjugated
FUBP1
DNA helicase V, FBP, FUBP, FUBP1, FUSE binding protein 1, hDH V
Rabbit
Antigen Affinity Chromatography
RUO
51886, 8880
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q91WJ8, Q96AE4
Fubp1
FUBP1 Fusion Protein Ag13149
20 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions