missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16567-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
FUT6 is a Golgi stack membrane protein that is involved in the creation of sialyl-Lewis X, an E-selectin ligand.Spécification
FUT6 | |
Polyclonal | |
Unconjugated | |
FUT6 | |
Alpha (1,3) fucosyltransferase, FCT3A, FT1A, Fuc TVI, Fucosyltransferase 6, Fucosyltransferase VI, FucT VI, FUT6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2528 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51993 | |
FUT6 | |
FUT6 Fusion Protein Ag9847 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |