missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD1 / GAD67 (GABAergic Neuronal Marker) Mouse, Clone: GAD1/2391, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 2571-MSM1-P0
Informations supplémentaires : Poids : 0.01000kg
Spécification
GAD1 / GAD67 (GABAergic Neuronal Marker) | |
Cytoplasmic | |
GAD1/2391 | |
Unconjugated | |
Mouse | |
67kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2571 | |
IgG1 κ |
ELISA, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human GAD1 (GAD67) protein fragment (apprx. aa 72-135) | |
K-562 or HEK293 Cells. Pancreas | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |