missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GAK (Human) Recombinant Protein
Human GAK full-length ORF ( AAH08668, 24 a.a. - 416 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00002580-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: DQSDFVGQTVELGELRLLLASPAPPLSVQSTPRGGPPAAADPFGPLLPSSGNNSQPCSNPDLFGEFLNSDSVTVPPSFPSAHSSPPPSCSADFLHLGDLPGEPSKMTASSSNPDLLGGWAAWTETAASAVAPTPATEGPLFSPGGQPAPCGSQASWTKSQNPDPFADLGDLSSGLQGSPAGFPPGGFIPKTATTPKGSSSWQTSRPPAQGASWPPQAKPPPKACTQPRPNYASNFSVIGAREERGVRAPSFAQKPKVSENDFEDLLSNQGFSSRSDKKGPKTIAEMRKQDLAKDTDPLKLKLLDWIEGKERNIRALLSTLHTVLWDGESRWTPVGMADLVAPEQVKKHYRRAVLAVHPDKAAGQPYEQHAKMIFMELNDAWSEFENQGSRPLF
Spécification
AAH08668 | |
cyclin G associated kinase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAK | |
GST |
2580 | |
Wheat germ expression system | |
10 μg | |
FLJ16629, FLJ40395, MGC99654 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |