missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15303-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Guanylate binding protein expression is induced by interferon. Guanylate binding proteins are characterized by their ability to specifically bind guanine nucleotides (GMP, GDP, and GTP) and are distinguished from the GTP-binding proteins by the presence of 2 binding motifs rather than 3.Spécification
GBP1 | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32455 | |
GBP 1, GBP1, GTP binding protein 1, HuGBP 1 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
2633 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
GBP1 | |
Liquid | |
GBP1 | |
GBP1 Fusion Protein Ag7562 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |