Learn More
Alfa Aesar™ Dihydrochloride GBR 12909
Inhibiteur du recaptage de dopamine
Marque: Alfa Aesar™ J60684.MF
Code nomenclature Nacres: NA.21
Informations supplémentaires : CAS : 67469-78-7 Poids : 0.00050kg
Quantité. | 50mg |
---|
Identifiants chimiques
67469-78-7 | |
523.49 | |
MIBSKSYCRFWIRU-UHFFFAOYSA-N | |
104920 | |
1-[2-[bis(4-fluorophényl)méthoxy]éthyl]-4-(3-phénylpropyl)pipérazine ;dichlorhydrate |
C28H34Cl2F2N2O | |
MFCD00055193 | |
vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride | |
CHEBI:64086 | |
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl |
Spécification
GBR 12909 dihydrochloride | |
50mg | |
C28H34Cl2F2N2O | |
vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride | |
MIBSKSYCRFWIRU-UHFFFAOYSA-N | |
1-[2-[bis(4-fluorophényl)méthoxy]éthyl]-4-(3-phénylpropyl)pipérazine ;dichlorhydrate | |
104920 | |
523.49 |
Blanc | |
67469-78-7 | |
MFCD00055193 | |
Soluble in DMSO to 50mM; Soluble to 5mM in water with warming. | |
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl | |
523.49 | |
CHEBI:64086 | |
Poudre |