Learn More
GE Healthcare Kit pour la prolifération cellulaire, kit et composants pour test ELISA BioTrak™
Provides a precise, fast and simple colorimetric assay to quantitate proliferating cells cultured in a 96 well microplate
Marque:  GE Healthcare RPN250
Code nomenclature Nacres: NA.32
Informations supplémentaires : Poids : 0.50000kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Code produit. 10095754
  • 1018.00€ / 960 pièces
Expédition estimée
22-04-2020
Ajouter au panier
Description
Description
  • Recommended cell number: 10e6-10e7 cells/mL (Cell proliferation kit) 10e2 cells/well-10e5 cells/well (Biotrak ELISA kit)
  • Required time: 2-3 hour post-labeling (labeling 1-2 hours)
  • Antibody and labeling reagent available separately for ELISA and ICC systems
  • Suitable for in vivo and in vitro tissue labeling in addition to cultured cells (ELISA format)
  • Non radioactive protocols provide sensitivity comparable to autoradiographic methods
  • Cell proliferation kit avoids the need to chemically denature DNA. Cellular morphology is preserved allowing multiple labeling experiments
  • 5-fluoro-2'-deoxyuridine is used to inhibit thymidilate synthetase increasing BrdU incorporation
When ordering the Cell Proliferation Kit (RPN20), you must also order DAB REAGENT OF RPN20/RPN1174 (RPNK8888)
Spécification
Spécification
102 to 105 Cells/well
10 x 96 Tests
≥ 10 2 cells/well
Immunocytochemistry
Suspension or adherent cells
2- 8°C
Vidéos
Sécurité et traitement
Sécurité et traitement
Mention d'avertissement
  • Dangers
Catégorie de danger
  • Liquide inflammable Catégorie 2
  • Corrosion cutanée/irritation cutanée Catégorie 1
Documents