missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24674-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
GGTL3 is an enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. GGTL3 consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.This gene is a member of a gene family that encodes enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. The protein encoded by this gene consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.Spécification
GGT7 | |
Polyclonal | |
Unconjugated | |
GGT7 | |
D20S101, gamma glutamyltransferase 7, Gamma glutamyltranspeptidase 7, GGT 7, GGT4, GGT7, GGTL3, GGTL5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
156275, 207182, 2686 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99JP7, Q99MZ4, Q9UJ14 | |
GGT7 | |
GGT7 Fusion Protein Ag20234 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |