missing translation for 'onlineSavingsMsg'
Learn More

GNL1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14078-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802564

  • 329.85€ / 150µL

Date d’expédition estimée: 22-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Spécification

Spécification

GNL1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P36915
GNL1, GTP binding protein HSR1, HSR1
Rabbit
IgG
150 μL
-20°C
Primary
2794
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GNL1
Liquid
GNL1
GNL1 Fusion Protein Ag5203
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents
Certificats