missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNPNAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16282-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Glucosamine-6 phosphate-N-acetyl Transferase (GNPNAT1) is a key enzyme in the hexosamine biosynthetic pathway. It converts D-glucosamine 6-phosphate to N-acetyl-D-glucosamine 6-phosphate, the acetylation step allowing carbon and nitrogen enter into the hexosamine biosynthetic pathway. Recently inhibition of GNPNAT1 has been reported to promote proliferation and aggressiveness of castration-resistant prostate cancer. (27194471).Spécification
GNPNAT1 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96EK6, Q9JK38 | |
GNA1, GNPNAT, GNPNAT1, Gpnat1, Phosphoglucosamine acetylase | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
498486, 54342, 64841 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
GNPNAT1 | |
Liquid | |
Gnpnat1 | |
GNPNAT1 Fusion Protein Ag9375 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |