Learn More
GPR161 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13398-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The 70 kDa band detected by this antibody probably represents a modified version of GPR161.
Upon ligand binding, G protein-coupled receptors, such as GPR161, activate cytoplasmic G proteins (see GNAS, MIM 139320), allowing the receptors to transduce extracellular signals across the plasma membrane into the cell.Spécification
GPR161 | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2RPY5, Q8N6U8 | |
G protein coupled receptor 161, G protein coupled receptor RE2, GPR161, RE2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
23432, 240888, 289180 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
GPR161 | |
Liquid | |
Gpr161 | |
GPR161 Fusion Protein Ag4221 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Rat, Human, Mouse |