missing translation for 'onlineSavingsMsg'
Learn More

GRK2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13990-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802454

  • 399.00€ / 150µL
Expédition estimée: 29-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The product of this gene phosphorylates the beta-2-adrenergic receptor and appears to mediate agonist-specific desensitization observed at high agonist concentrations. This protein is an ubiquitous cytosolic enzyme that specifically phosphorylates the activated form of the beta-adrenergic and related G-protein-coupled receptors. Abnormal coupling of beta-adrenergic receptor to G protein is involved in the pathogenesis of the failing heart.
Spécification

Spécification

GRK2
Polyclonal
Unconjugated
GRK2
ADRBK1, BARK, BARK1, Beta ARK 1, BETA ARK1, GRK2
Rabbit
Antigen Affinity Chromatography
RUO
156
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P25098
GRK2
GRK2 Fusion Protein Ag5071
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions