missing translation for 'onlineSavingsMsg'
Learn More

Growth hormone Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00€ - 399.00€

Spécification

Antigène Growth hormone
Concentration 0.27 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16805375
Afficher les documents Détails de la promotion
Proteintech
27079-1-AP-20UL
20 μL
140.00€
20µL
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16895365
Afficher les documents Détails de la promotion
Proteintech
27079-1-AP-150UL
150 μL
399.00€
150µL
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.
Spécification

Spécification

Growth hormone
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
P01241
2688
Growth hormone Fusion Protein Ag22841
Primary
-20°C
GH1
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
GH, GH N, GH1, GHN, Growth hormone, growth hormone 1, hGH N, Pituitary growth hormone, Somatotropin
GH1
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications