missing translation for 'onlineSavingsMsg'
Learn More

GTF2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00€ - 399.00€

Spécification

Antigène GTF2B
Concentration 0.4 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16806104
Afficher les documents Détails de la promotion
Proteintech
16467-1-AP-20UL
20 μL
140.00€
20µL
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16896094
Afficher les documents Détails de la promotion
Proteintech
16467-1-AP-150UL
150 μL
399.00€
150µL
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Ubiquitin is a 76 amino acid highly conserved eukaryotic polypeptide that selectively marks cellular proteins for proteolytic degradation by the 26S proteasome. The process of target selection, covalent attachment and shuttle to the 26S proteasome is a vital means of regulating the concentrations of key regulatory proteins in the cell by limiting their lifespans. Polyubiquitination is a common feature of this modification. Serial steps for modification include the activation of ubiquitin, an ATP-dependent formation of a thioester bond between ubiquitin and the enzyme E1, transfer by transacylation of ubiquitin from E1 to the ubiquitin conjugating enzyme E2, and covalent linkage to the target protein directly by E2 or via E3 ligase enzyme. Deubiquitination enzymes also exist to reverse the marking of protein substrates. Posttranslational tagging by Ub is involved in a multitude of cellular processes, including the cell cycle, cell growth and differentiation, embryogenesis, apoptosis, signal transduction, DNA repair, regulation of transcription and DNA replication, transmembrane transport, stress responses, the immune response, and nervous system functions.
Spécification

Spécification

GTF2B
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P62915, P62916, Q00403
229906, 2959, 81673
GTF2B Fusion Protein Ag9547
Primary
-20°C
GTF2B
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
GTF2B, S300 II, TF2B, TFIIB
GTF2B
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications