Learn More
HBS1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10359-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody specifically recognizes the 75 kDa human HBS1L protein.
This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.Spécification
HBS1L | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q69ZS7, Q6AXM7, Q9Y450 | |
EF 1a, ERFS, HBS1, HBS1 like (S. cerevisiae), HBS1 like protein, HBS1L, HSPC276, KIAA1038 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
10767, 293408, 56422 |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
HBS1L | |
Liquid | |
Hbs1l | |
HBS1L Fusion Protein Ag0388 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Rat, Mouse, Human |