missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDDC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16632-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The function of this protein remains unknown.Spécification
HDDC2 | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3SXD3, Q7Z4H3 | |
C6orf74, CGI 130, HD domain containing 2, HD domain containing protein 2, HDDC2, NS5ATP2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
361462, 51020, 69692 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
HDDC2 | |
Liquid | |
HDDC2 | |
HDDC2 Fusion Protein Ag9971 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |