missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemopexin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15736-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.Spécification
Hemopexin | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02790 | |
hemopexin, HPX | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
3263 |
Western Blot | |
Unconjugated | |
HPX | |
Liquid | |
HPX | |
Hemopexin Fusion Protein Ag8533 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |