missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepsin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51060-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Hepsin is a cell surface serine protease.Spécification
Hepsin | |
Polyclonal | |
Unconjugated | |
HPN | |
AA408042, AI788568, hepsin, Hpn | |
Rabbit | |
Protein A | |
RUO | |
15451, 3249 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35453, P05981 | |
Hpn | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |