missing translation for 'onlineSavingsMsg'
Learn More

HIF3A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27650-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805765

  • 112.71€ / 20µL

Expédition estimée
09-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.
Spécification

Spécification

HIF3A
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y2N7
bHLHe17, HIF 3 alpha, HIF 3A, HIF 3A2, HIF 3A4, HIF3 alpha, HIF3 alpha 1, HIF3A, Inhibitory PAS domain protein, IPAS, Member of PAS protein 7, MOP7, PASD7
Rabbit
IgG
20 μL
-20°C
Primary
64344
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
HIF3A
Liquid
HIF3A
HIF3A Fusion Protein Ag26574
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents