missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HKR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | HKR1 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16843804
|
Proteintech
14902-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16833804
|
Proteintech
14902-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. HKR1, also known as Krueppel-related zinc finger protein 1 or zinc finger protein 875, is a 659 amino acid nuclear protein that is thought to play a role in transcriptional regulation. Existing as two alternatively spliced isoforms, HKR1 is a member of the Krueppel C2H2-type zinc-finger protein family and contains thirteen C2H2-type zinc fingers and one KRAB domain. The gene encoding HKR1 maps to human chromosome 19q13.12.Spécification
HKR1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P10072 | |
284459 | |
HKR1 Fusion Protein Ag6694 | |
Primary | |
-20°C | |
HKR1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GLI Kruppel family member HKR1, HKR1, Protein HKR1 | |
HKR1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |